missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide-d6, 100% (Isotopic), Thermo Scientific Chemicals

Product Code. p-7051938
Click to view available options
Quantity:
10 Ea.
Unit Size:
Pack of 10
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11339977

Brand: Thermo Scientific Alfa Aesar 042285.KV

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Deuterated DMSO is a common solvent used in NMR spectroscopy. In combination with pyridine-d5, it plays an important role for solution-state 2D NMR of ball-milled plant cell wall gel. Low-energy collisions of ionized acetone-d6, DMSO-d6 and pyridine-d5 with alkanethiolate is used to prepare self-assembled monolayer surfaces of gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 1.18 g/mL
Flash Point 88°C (190°F)
Odor Characteristic
Packaging Prescored Ampule
Refractive Index 1.476
Sensitivity Hygroscopic
Quantity 10 Ea.
Melting Point 20°C
Boiling Point 190°C
Solubility Information Miscible with water and diethyl ether.
Formula Weight 84.17
Percent Purity 100%
Chemical Name or Material Dimethyl sulfoxide-d6, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.