missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals

Product Code. 15488089
Click to view available options
Quantity:
50 mL
100 mL
250 mL

DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.

Solubility
Soluble in water, methanol, acetone, ether, benzene, chloroform.

Notes
Submicron filtered. Store at Room Temperature (15-30°C), desiccate. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Density 1.101 g/mL
Boiling Point 189°C
Quantity 50 mL
Flash Point 87°C (189°F)
Odor Mild
Physical Form Liquid
Refractive Index 1.479
Beilstein 506008
Solubility Information Soluble in water,methanol,acetone,ether,benzene,chloroform.
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Chemical Name or Material Dimethyl sulfoxide, Bioreagent
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'