missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, 99.97%, MP Biomedicals™

Product Code. 11446521 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
This item is not returnable. View return policy

Product Code. 11446521

Brand: MP Biomedicals 0219481950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Color Colorless
Density 1.10g/mL
Quantity 50 mL
Physical Form Liquid To Solid
Assay Percent Range 0.9997
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Chemical Name or Material Dimethyl sulfoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.