missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, ≥99.9%, Solstice

Product Code. 15685630
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
6 x 500 mL
4 x 4 L
6 x 1 L
Unit Size:
1 L
100mL
2.5 L
4 L
500mL
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15685630 500 mL 500mL
15645630 100 mL 100mL
15655630 1 L 1 L
15665630 2.5 L 2.5 L
15605640 6 x 500 mL Pack of 6
15658410 4 L 4 L
15695630 6 x 1 L Pack of 6
15675630 4 x 4 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15685630 Supplier Honeywell-Riedel-de Haen Supplier No. 472301500ML
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
This item is not returnable. View return policy

ACS Reagent,≥99.9%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Color Colorless
Density 1.10g/mL
Quantity 500 mL
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
IUPAC Name methanesulfinylmethane
Formula Weight 78.13
Percent Purity ≥99.9%
Grade ACS
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.