missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide (13C2, 99%), TRC

Product Code. 30383572
Change view
Click to view available options
Quantity:
20 mg
Unit Size:
20mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383572 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383572 Supplier Toronto Research Chemicals Supplier No. TRCD48074020MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl Sulfoxide-13C2
CAS 136321-15-8
Molecular Formula 13C2H6OS
Quantity 20 mg
Synonym DMSO-13C2, Dimethyl Sulfoxide (13C2), 13C Labeled dimethyl sulfoxide, Methyl sulfoxide-13C2
InChI Formula InChI=1S/C2H6OS/c1-4(2)3/h1-2H3/i1+1,2+1
SMILES [13CH3]S(=O)[13CH3]
IUPAC Name ((methyl-13C)sulfinyl)methane-13C; Methane-13C, sulfinylbis- (9CI)
Molecular Weight (g/mol) 80.11
Formula Weight 80.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.