Learn More
Dimethyl sulfone, 98%
Description
Dimethyl sulfone is used as a high-temperature solvent for both inorganic and organic substances. It acts as an intermediate as well as medium in organic synthesis. It plays an important role as a carbon source by the airborne bacteria Afipia. It is also employed as an extraction solvent for the separation of aromatic compounds like benzene, toluene and xylenes from aliphatic hydrocarbons. Further, it is used in the polymerization process. It finds application in pharmaceuticals, agrochemicals, paint and coating materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl sulfone is used as a high-temperature solvent for both inorganic and organic substances. It acts as an intermediate as well as medium in organic synthesis. It plays an important role as a carbon source by the airborne bacteria Afipia. It is also employed as an extraction solvent for the separation of aromatic compounds like benzene, toluene and xylenes from aliphatic hydrocarbons. Further, it is used in the polymerization process. It finds application in pharmaceuticals, agrochemicals, paint and coating materials.
Solubility
Soluble in water, methanol, ethanol and acetone.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 108°C to 111°C |
| Density | 1.45 |
| Boiling Point | 238°C |
| Flash Point | 143°C (289°F) |
| Odor | Odorless |
| Quantity | 1000 g |
| Beilstein | 1737717 |
| Merck Index | 14,3258 |
| Solubility Information | Soluble in water,methanol,ethanol and acetone. |
| Formula Weight | 94.13 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.