missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfite, 99%

Product Code. 10675002 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10675002 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10675002 Supplier Thermo Scientific Acros Supplier No. 148751000
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-42-2 | C2H6O3S | 110.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Specifications

CAS 616-42-2
Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 126°C to 127°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C2H6O3S
Linear Formula (CH3O)2SO
Refractive Index 1.4085 to 1.4105
MDL Number MFCD00008415
Quantity 100 mL
Beilstein 01, 282
Fieser 01,296
Specific Gravity 1.29
Synonym methyl sulfite, sulfurous acid, dimethyl ester, dimethyl sulphite, dimethoxy sulfoxide, dimethylsulfit, unii-9jfa40s66p, ch3o 2so, sulphurous acid dimethyl ester, dimethyl ester of sulfurous acid, dimethylsulfite
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmf,slightly soluble in nonpolar solvents
InChI Key BDUPRNVPXOHWIL-UHFFFAOYSA-N
SMILES COS(=O)OC
IUPAC Name dimethyl sulfite
Molecular Weight (g/mol) 110.13
PubChem CID 69223
ChEBI CHEBI:48858
Formula Weight 110.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl sulfite, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl sulfite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.