missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfate, TRC

Product Code. 30004650
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004650 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30004650 Supplier Toronto Research Chemicals Supplier No. TRCD47930510G
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 77-78-1
Molecular Formula C2H6O4S
Quantity 10 g
Synonym Sulfuric Acid Dimethyl Ester, Dimethyl Sulphate, Methyl Sulfate, NSC 56194, DMS
InChI Formula InChI=1S/C2H6O4S/c1-5-7(3,4)6-2/h1-2H3
SMILES COS(=O)(=O)OC
Molecular Weight (g/mol) 126.13
Formula Weight 125.9987
Percent Purity >95%
Chemical Name or Material Dimethyl Sulfate
Grade GC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.