missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Succinate, TRC

Product Code. 30380968
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380968 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380968 Supplier Toronto Research Chemicals Supplier No. TRCD472815250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Succinic acid dimethyl ester
CAS 106-65-0
Molecular Formula C6 H10 O4
Quantity 250 g
Synonym Butanedioic acid, 1,4-dimethyl ester (ACI), Butanedioic acid, dimethyl ester (9CI), Succinic acid, dimethyl ester (6CI, 8CI), DBE 4, Dimethyl 1,4-butanedioate, Dimethyl butanedioate, Dimethyl succinate, Methyl succinate, NSC 52209, Provichem 2511, Proviron 2511ECO, Rhodiasolv RDEP, Succinic acid-dimethyl ester
InChI Formula InChI=1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3
SMILES COC(=O)CCC(=O)OC
IUPAC Name dimethyl butanedioate
Molecular Weight (g/mol) 146.14
Formula Weight 146.0579
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.