missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Succinate, TRC

Product Code. 30370932
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370932 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370932 Supplier Toronto Research Chemicals Supplier No. TRCD472815100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Succinic acid dimethyl ester
CAS 106-65-0
Molecular Formula C6 H10 O4
Quantity 100 g
Synonym Butanedioic acid, 1,4-dimethyl ester (ACI), Butanedioic acid, dimethyl ester (9CI), Succinic acid, dimethyl ester (6CI, 8CI), DBE 4, Dimethyl 1,4-butanedioate, Dimethyl butanedioate, Dimethyl succinate, Methyl succinate, NSC 52209, Provichem 2511, Proviron 2511ECO, Rhodiasolv RDEP, Succinic acid-dimethyl ester
InChI Formula InChI=1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3
SMILES COC(=O)CCC(=O)OC
IUPAC Name dimethyl butanedioate
Molecular Weight (g/mol) 146.14
Formula Weight 146.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.