missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Succinate, TRC
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| Chemical Name or Material | Succinic acid dimethyl ester |
| CAS | 106-65-0 |
| Molecular Formula | C6 H10 O4 |
| Quantity | 500 g |
| Synonym | Butanedioic acid, 1,4-dimethyl ester (ACI), Butanedioic acid, dimethyl ester (9CI), Succinic acid, dimethyl ester (6CI, 8CI), DBE 4, Dimethyl 1,4-butanedioate, Dimethyl butanedioate, Dimethyl succinate, Methyl succinate, NSC 52209, Provichem 2511, Proviron 2511ECO, Rhodiasolv RDEP, Succinic acid-dimethyl ester |
| InChI Formula | InChI=1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3 |
| SMILES | COC(=O)CCC(=O)OC |
| IUPAC Name | dimethyl butanedioate |
| Molecular Weight (g/mol) | 146.14 |
| Formula Weight | 146.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction