missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl selenide

Product Code. 11374789
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374789 25 g 25g
11356319 1 g 1g
11364789 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11374789 Supplier Thermo Scientific Alfa Aesar Supplier No. 068116.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-79-3 | C2H6Se | 109.041 g/mol

Used as primary and secondary intermediate and also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-79-3
Molecular Formula C2H6Se
Molecular Weight (g/mol) 109.041
MDL Number MFCD00014848
InChI Key RVIXKDRPFPUUOO-UHFFFAOYSA-N
Synonym dimethylselenide, dimethylselenium, dimethyl selenide, methyl selenide, methyl selenium, methane, selenobis, selenium dimethyl, selenide, dimethyl, ch3 2se, unii-yk0r6jkt6h
PubChem CID 11648
ChEBI CHEBI:4610
IUPAC Name methylselanylmethane
SMILES C[Se]C

Specifications

Color Colorless
Density 1.41 g/mL
Boiling Point 56°C to 58°C
Odor Unpleasant
Linear Formula (CH3)2Se
Quantity 25 g
UN Number UN3282
Beilstein 1696848
Sensitivity Air and light sensitive
Solubility Information Very soluble in ethyl ether and ethyl alcohol.
Formula Weight 109.03
Physical Form Liquid
Chemical Name or Material Dimethyl selenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.