missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (S)-(-)-Malate, TRC

Product Code. 30370789
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370789 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370789 Supplier Toronto Research Chemicals Supplier No. TRCD4745755G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 617-55-0
Molecular Formula C6 H10 O5
Quantity 5 g
Synonym Butanedioic acid, 2-hydroxy-, 1,4-dimethyl ester, (2S)-, Butanedioic acid, hydroxy-, dimethyl ester, (2S)- (9CI), Butanedioic acid, hydroxy-, dimethyl ester, (S)-, Malic acid, dimethyl ester, L- (8CI), (-)-Dimethyl malate, (2S)-Hydroxysuccinic acid dimethyl ester, (S)-(-)-Dimethyl malate, (S)-Dimethyl 2-hydroxysuccinate, (S)-Malic acid dimethyl ester, 1,4-Dimethyl (2S)-2-hydroxybutanedioate, Dimethyl (2S)-2-hydroxysuccinate, Dimethyl (S)-(-)-2-hydroxysuccinate, Dimethyl (S)-2-hydroxybutanedioate, Dimethyl (S)-malate, Dimethyl L-malate, L-Malic acid dimethyl ester
InChI Formula InChI=1S/C6H10O5/c1-10-5(8)3-4(7)6(9)11-2/h4,7H,3H2,1-2H3/t4-/m0/s1
SMILES COC(=O)C[C@H](O)C(=O)OC
IUPAC Name dimethyl (2S)-2-hydroxybutanedioate
Molecular Weight (g/mol) 162.14
Formula Weight 162.0528
Chemical Name or Material Dimethyl (S)-(-)-Malate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.