missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (S)-(-)-malate, 98%

Product Code. 11905091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11905091 5 g Glass bottle 5g
11925091 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905091 Supplier Thermo Scientific Acros Supplier No. 443240050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 617-55-0 | C6H10O5 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-55-0
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00066215
InChI Key YSEKNCXYRGKTBJ-UHFFFAOYNA-N
Synonym s-dimethyl 2-hydroxysuccinate, dimethyl s---malate, dimethyl l-malate, l---malic acid dimethyl ester, 1,4-dimethyl 2s-2-hydroxybutanedioate, butanedioic acid, hydroxy-, dimethyl ester, 2s, dimethyl l---malate, pubchem6778, di-methyl s-malate, dimethyl s--malate
PubChem CID 10285815
IUPAC Name dimethyl (2S)-2-hydroxybutanedioate
SMILES COC(=O)CC(O)C(=O)OC

Specifications

Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 91.0°C to 93.0°C (1.0 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H3COOCCH(OH)CH2COOCH3
Refractive Index 1.4366
Quantity 5 g
Specific Gravity 1.23
Specific Rotation − 6.70
Solubility Information Solubility in water: in water:(20 °C) insoluble.
Formula Weight 162.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl (S)-(-)-malate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl (S)-(-)-malate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.