missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl pyridine-2,5-dicarboxylate, 97%

Product Code. 11483477
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11483477

Brand: Thermo Scientific Alfa Aesar A17250.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 881-86-7 | C9H9NO4 | 195.174 g/mol

It is the intermediate in the synthesis of potent antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the intermediate in the synthesis of potent antitumor agents.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents, reducing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881-86-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00034767
InChI Key TUGSJNQAIMFEDY-UHFFFAOYSA-N
Synonym dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
PubChem CID 234911
IUPAC Name dimethyl pyridine-2,5-dicarboxylate
SMILES COC(=O)C1=CN=C(C=C1)C(=O)OC

Specifications

Melting Point 160°C to 163°C
Quantity 100 g
Beilstein 172251
Solubility Information Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.
Formula Weight 195.17
Percent Purity ≥98%
Chemical Name or Material Dimethyl pyridine-2,5-dicarboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.