missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 234.21 |
Percent Purity | 97% |
Chemical Name or Material | Dimethyl pyrazolo[1, 5-a]pyridine-2, 3-dicarboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'