missing translation for 'onlineSavingsMsg'
Få mere at vide

Dimethyl Phthalate, Extra Pure, SLR, Fisher Chemical™

Artikelnummer. 10364980 Shop alle Fisher Chemical produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
500 mL
2.5 L
Pakningsstørrelse:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
10364980 500 mL 500mL
11418213 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10364980 Leverandør Fisher Chemical Leverandørnr. D/4050/08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008425
InChI Key NIQCNGHVCWTJSM-UHFFFAOYSA-N
Synonym dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
PubChem CID 8554
ChEBI CHEBI:4609
IUPAC Name dimethyl benzene-1,2-dicarboxylate
SMILES COC(=O)C1=CC=CC=C1C(=O)OC

Specifications

Melting Point 2°C
Color Colorless
Boiling Point 282°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 6.69
Formula Weight 194.19g/mol
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.