missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl phthalate, 99%

Product Code. 11412387
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412387 500 g 500g
11374479 250 g 250g
11422387 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412387 Supplier Thermo Scientific Alfa Aesar Supplier No. A14253.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131-11-3 | C10H10O4 | 194.19 g/mol

Dimethyl phthalate is used as a plasticizer, solvent, rubber coating agent, molding powder, finishing agent and intermediate. Further, it is used in solid rocket propellants, safety glass, lacquer coatings, adhesives and sealants, toner and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phthalate is used as a plasticizer, solvent, rubber coating agent, molding powder, finishing agent and intermediate. Further, it is used in solid rocket propellants, safety glass, lacquer coatings, adhesives and sealants, toner and colorant products.

Solubility
Miscible with alcohol, ethanol, ether, benzene, chloroform, esters, ketones, organic solvents and chlorinated hydrocarbons. Slightly miscible with carbon tetrachloride. Immiscible with water, petroleum ether and paraffin hydrocarbons.

Notes
Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008425
InChI Key NIQCNGHVCWTJSM-UHFFFAOYSA-N
Synonym dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
PubChem CID 8554
ChEBI CHEBI:4609
SMILES COC(=O)C1=CC=CC=C1C(=O)OC

Specifications

Melting Point ∽5°C
Density 1.190
Boiling Point 283°C to 284°C
Flash Point 146°C (294°F)
Assay Percent Range 99%
Refractive Index 1.515
Quantity 500 g
Beilstein 1911460
Merck Index 14,3255
Solubility Information Miscible with alcohol,ethanol,ether,benzene,chloroform,esters,ketones,organic solvents and chlorinated hydrocarbons. Slightly miscible with carbon tetrachloride. Immiscible with water,petroleum ether and paraffin hydrocarbons.
IUPAC Name dimethyl benzene-1,2-dicarboxylate
Formula Weight 194.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl Phthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.