missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phthalate, 99%, Thermo Scientific Chemicals

Catalog Number p-3736336
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy
131-11-3
C10H10O4
194.19
MFCD00008425
NIQCNGHVCWTJSM-UHFFFAOYSA-N
dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
8554
CHEBI:4609
dimethyl benzene-1,2-dicarboxylate
COC(=O)C1=CC=CC=C1C(=O)OC
This item is not returnable. View return policy
131-11-3
C10H10O4
194.19
MFCD00008425
NIQCNGHVCWTJSM-UHFFFAOYSA-N
dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
8554
CHEBI:4609
dimethyl benzene-1,2-dicarboxylate
COC(=O)C1=CC=CC=C1C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.1900g/mL
Boiling Point 282°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5140 to 1.516
Quantity 250 mL
Beilstein 09, 797
Merck Index 15, 3279
Specific Gravity 1.19
Solubility Information Solubility in water: negligible. Other solubilities: miscible with organic solvents,insoluble in paraffinic hydrocarbons,slightly soluble in mineral oil
Viscosity 17 mPa.s (20°C)
Formula Weight 194.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl phthalate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'