missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl phthalate 5000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30319667
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319667 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319667 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011090ME
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-methyl ester
CAS 131-11-3
Molecular Formula C10 H10 O4
Quantity 1 mL
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzenedicarboxylic acid, dimethyl ester (9CI), Phthalic acid, dimethyl ester (6CI,8CI), Avolin, DMF (insect repellent), DMP, Dimethyl 1,2-benzenedicarboxylate, Dimethyl o-phthalate, Dimethyl phthalate, Fermine, Metox 50, Mipax, NSC 15398, NTM, Palatinol M, Repeftal, Solvanom, Solvarone, Unimoll DM
InChI Formula InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3
SMILES COC(=O)c1ccccc1C(=O)OC
IUPAC Name dimethyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 194.18
Formula Weight 194.0579 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.