missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phthalate-13C2, TRC

Product Code. 30383862
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383862 50 mg
30381549 100 mg
30387672 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383862 Supplier Toronto Research Chemicals Supplier No. TRCD47797750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346598-73-9
Molecular Formula C813C2H10O4
Molecular Weight (g/mol) 196.17
Synonym 1,2-Benzenedicarboxylic Acid 1,2-Dimethyl-13C2 Ester, Avolin-13C2, DMF-13C2, DMP-13C2, Dimethyl-13C2 1,2-Benzenedicarboxylate, Dimethyl-13C2 o-Phthalate, Dimethyl-13C2 Phthalate, Fermine-13C2, Mipax-13C2, NSC 15398-13C2, NTM-13C2, Palatinol M-13C2, Repeftal-13C2, Solvanom-13C2, Solvarone-13C2, Unimoll DM-13C2
SMILES [13CH3]OC(=O)c1ccccc1C(=O)O[13CH3]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i1+1,2+1
Formula Weight 196.0646
Chemical Name or Material Dimethyl Phthalate-13C2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.