missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl oxazole-4,5-dicarboxylate, 99%, Thermo Scientific™

Product Code. 11395055
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395055 1 g 1g
11385055 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395055 Supplier Thermo Scientific Alfa Aesar Supplier No. H30009.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The teroxazole has been synthesized from dimethyl oxazole-4,5-dicarboxylate and lithiobenzyl isocyanide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72030-81-0
Molecular Formula C7H7NO5
Molecular Weight (g/mol) 185.135
MDL Number MFCD05863258
InChI Key LYRDBSFJJDATFO-UHFFFAOYSA-N
Synonym dimethyl oxazole-4,5-dicarboxylate, 4,5-dimethyl 1,3-oxazole-4,5-dicarboxylate, methyl 5-methoxycarbonyl-1,3-oxazole-4-carboxylate
PubChem CID 819934
IUPAC Name dimethyl 1,3-oxazole-4,5-dicarboxylate
SMILES COC(=O)C1=C(OC=N1)C(=O)OC

Specifications

Melting Point 74°C to 76°C
Boiling Point 150°C (12 mmHg)
Quantity 1 g
Formula Weight 185.14
Percent Purity 99%
Chemical Name or Material Dimethyl oxazole-4,5-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.