missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Oxalate, TRC

Product Code. 30388022
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388022 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388022 Supplier Toronto Research Chemicals Supplier No. TRCD4763451G
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxalic acid methyl ester
CAS 553-90-2
Molecular Formula C4 H6 O4
Quantity 1 g
Synonym Oxalic acid, bis-methyl ester, Ethanedioic acid, 1,2-dimethyl ester (ACI), Ethanedioic acid, dimethyl ester (9CI), Oxalic acid, dimethyl ester (6CI, 8CI), Dimethyl oxalate, Methyl oxalate, NSC 9374
InChI Formula InChI=1S/C4H6O4/c1-7-3(5)4(6)8-2/h1-2H3
SMILES COC(=O)C(=O)OC
IUPAC Name dimethyl oxalate
Molecular Weight (g/mol) 118.09
Formula Weight 118.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.