missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl methoxymethylenemalonate, 98+%

Product Code. 10511131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10511131 250 g Glass bottle 250g
10023270 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10511131 Supplier Thermo Scientific Acros Supplier No. 269562500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22398-14-7 | C7H10O5 | 174.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22398-14-7
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.16
MDL Number MFCD00038225
InChI Key RHFZTBSULNJWEI-UHFFFAOYSA-N
Synonym dimethyl methoxymethylenemalonate, dimethyl 2-methoxymethylene malonate, 1,3-dimethyl 2-methoxymethylidene propanedioate, methoxymethylenemalonic acid dimethyl ester, dimethyl methoxymethylene malonate, acmc-1cq9e, dimethylmethoxymethylenemalonate, propanedioic acid, methoxymethylene-, dimethyl ester, dimethyl methoxymethylidenemalonate
PubChem CID 90770
IUPAC Name dimethyl 2-(methoxymethylidene)propanedioate
SMILES COC=C(C(=O)OC)C(=O)OC

Specifications

Melting Point 40°C to 44°C
Color White
Boiling Point 98°C (0.5 mmHg)
Flash Point 158°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH=C(COOCH3)2
Quantity 250 g
Solubility Information Solubility in water: may hydrolyse. Other solubilities: soluble in methanol 1260g/L (20 c)
Formula Weight 174.16
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material Dimethyl methoxymethylenemalonate, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl methoxymethylenemalonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.