missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl methoxymethylenemalonate, 98+%, Thermo Scientific™

Product Code. 11410268
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410268 250 g 250g
11400268 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410268 Supplier Thermo Scientific Alfa Aesar Supplier No. B23803.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22398-14-7
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.152
MDL Number MFCD00038225
InChI Key RHFZTBSULNJWEI-UHFFFAOYSA-N
Synonym dimethyl methoxymethylenemalonate, dimethyl 2-methoxymethylene malonate, 1,3-dimethyl 2-methoxymethylidene propanedioate, methoxymethylenemalonic acid dimethyl ester, dimethyl methoxymethylene malonate, acmc-1cq9e, dimethylmethoxymethylenemalonate, propanedioic acid, methoxymethylene-, dimethyl ester, dimethyl methoxymethylidenemalonate
PubChem CID 90770
IUPAC Name dimethyl 2-(methoxymethylidene)propanedioate
SMILES COC=C(C(=O)OC)C(=O)OC

Specifications

Melting Point 41°C to 45°C
Boiling Point 97°C to 99°C (0.5 mmHg)
Flash Point 158°C (316°F)
Odor Ester-like
Quantity 250 g
Formula Weight 174.16
Percent Purity ≥98%
Chemical Name or Material Dimethyl methoxymethylenemalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.