missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl malonate, 99+%, Thermo Scientific Chemicals

Catalog Number p-118288
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
108-59-8
C5H8O4
132.12
MFCD00008460
BEPAFCGSDWSTEL-UHFFFAOYSA-N
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
7943
dimethyl propanedioate
COC(=O)CC(=O)OC
This item is not returnable. View return policy
108-59-8
C5H8O4
132.12
MFCD00008460
BEPAFCGSDWSTEL-UHFFFAOYSA-N
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
7943
dimethyl propanedioate
COC(=O)CC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -62.0°C
Color Colorless
Density 1.1560g/mL
Boiling Point 180.0°C to 181.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH2CO2CH3
Refractive Index 1.4120 to 1.4140
Quantity 250 mL
Beilstein 02,572
Fieser 08,193; 13,34
Merck Index 15,6168
Specific Gravity 1.156
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol and ether
Formula Weight 132.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dimethyl malonate, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'