missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Maleate, TRC

Product Code. 30371390
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30371390

Brand: Toronto Research Chemicals TRCD47357010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maleic acid dimethyl ester
CAS 624-48-6
Molecular Formula C6 H8 O4
Quantity 10 g
Synonym Dimethyl Maleate, 2-Butenedioic acid (2Z)-, 1,4-dimethyl ester (ACI), 2-Butenedioic acid (2Z)-, dimethyl ester (9CI), 2-Butenedioic acid (Z)-, dimethyl ester (ZCI), Maleic acid, dimethyl ester (6CI, 8CI), (2Z)-2-Butenedioic acid dimethyl ester, (Z)-2-Butenedioic acid dimethyl ester, (Z)-Dimethyl 2-butenedioate, 1,4-Dimethyl (2Z)-but-2-enedioate, Dimethyl (Z)-but-2-enedioate, Dimethyl (Z)-butenedioate, Dimethyl cis-butenedioate, Dimethyl cis-ethenedicarboxylate, Dimethyl cis-ethylenedicarboxylate, Dimethyl maleate, Methyl maleate, Sipomer DMM
InChI Formula InChI=1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3-
SMILES COC(=O)\C=C/C(=O)OC
IUPAC Name dimethyl (Z)-but-2-enedioate
Molecular Weight (g/mol) 144.13
Formula Weight 144.0423
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.