missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl maleate, 96%

Product Code. 15730839 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
This item is not returnable. View return policy

Product Code. 15730839

Brand: Thermo Scientific Acros 218462500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-48-6 | C6H8O4 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-48-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00008459
InChI Key LDCRTTXIJACKKU-ARJAWSKDSA-N
Synonym dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z
PubChem CID 5271565
ChEBI CHEBI:35460
IUPAC Name dimethyl (Z)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point -17.5°C
Density 1.1500g/mL
Boiling Point 204.0°C to 205.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH=CHCO2CH3
Refractive Index 1.4410 to 1.4430
Quantity 250 mL
Beilstein 02,751
Specific Gravity 1.15
Solubility Information Solubility in water: 80g/L (25°C)
Viscosity 4 mPa.s (20°C)
Formula Weight 144.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Dimethyl maleate, 96%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.