missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl L-Glutamate Hydrochloride, Mikromol™

Product Code. 30050610
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050610 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050610 Supplier Mikromol Supplier No. MM1301.030025
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dimethyl L-Glutamate Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl L-Glutamate Hydrochloride
CAS 23150-65-4
Analyte or Component Names Dimethyl L-Glutamate Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C7 H13 N O4 . Cl H
Certifications/Compliance ISO 9001
Synonym L-Glutamic acid, 1,5-dimethyl ester, hydrochloride (1:1), Glutamic acid, dimethyl ester, hydrochloride, L- (7CI,8CI), L-Glutamic acid, dimethyl ester, hydrochloride (9CI), Dimethyl (S)-2-aminopentanedioate hydrochloride, Dimethyl L-glutamate hydrochloride, Dimethyl glutamate hydrochloride, Glutamic acid dimethyl ester hydrochloride, NSC 83254
InChI Formula InChI=1S/C7H13NO4.ClH/c1-11-6(9)4-3-5(8)7(10)12-2;/h5H,3-4,8H2,1-2H3;1H/t5-;/m0./s1
SMILES Cl.COC(=O)CC[C@H](N)C(=O)OC
IUPAC Name dimethyl (2S)-2-aminopentanedioate;hydrochloride
Molecular Weight (g/mol) 211.6434
Quantity 25 mg
Formula Weight 211.0611 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.