missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl L-Aspartate Hydrochloride, TRC

Product Code. 30370365
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370365 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370365 Supplier Toronto Research Chemicals Supplier No. TRCD4623451G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl L-Aspartate Hydrochloride
CAS 32213-95-9
Molecular Formula C6H11NO4.HCl
Quantity 1 g
Synonym L-Aspartic Acid Dimethyl Ester Hydrochloride, Dimethyl (S)-2-Aminosuccinate Hydrochloride, Dimethyl (S)-Aspartate Hydrochloride, Dimethyl Aspartate Hydrochloride, NSC 112494
InChI Formula InChI=1S/C6H11NO4.ClH/c1-10-5(8)3-4(7)6(9)11-2;/h4H,3,7H2,1-2H3;1H/t4-;/m0./s1
SMILES COC(C[C@H](N)C(OC)=O)=O.Cl
IUPAC Name dimethyl L-aspartate hydrogen chloride
Molecular Weight (g/mol) 197.62
Formula Weight 197.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.