missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Itaconate, TRC

Product Code. 30374941
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374941 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374941 Supplier Toronto Research Chemicals Supplier No. TRCD47420525G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Itaconic Acid-Dimethyl Ester
CAS 617-52-7
Molecular Formula C7 H10 O4
Quantity 25 g
Synonym Butanedioic acid, methylene-, dimethyl ester (9CI), Itaconic acid dimethyl ester (6CI), Succinic acid, methylene-, dimethyl ester (8CI), 1,4-Dimethyl 2-methylidenebutanedioate, 2-(Methylene)butanedioic dimethyl ester, 2-Methylene-succinic acid dimethyl ester, Dimethyl 2-methylenesuccinate, Dimethyl itaconate, Dimethyl methylenebutanedioate, Dimethyl methylenesuccinate, Methyl 4-methoxy-2-methylene-4-oxobutanoate, Methylenebutanedioic acid dimethyl ester, Methylenesuccinic acid dimethyl ester, NSC 9385
InChI Formula InChI=1S/C7H10O4/c1-5(7(9)11-3)4-6(8)10-2/h1,4H2,2-3H3
SMILES COC(=O)CC(=C)C(=O)OC
IUPAC Name dimethyl 2-methylidenebutanedioate
Molecular Weight (g/mol) 158.152
Formula Weight 158.058
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.