missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isophthalate, 99%

Product Code. 10164250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164250 2.5 kg Plastic bottle 2.5kg
10225860 25 g Plastic bottle 25g
10675822 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164250 Supplier Thermo Scientific Acros Supplier No. 186510025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1459-93-4 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1459-93-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008433
InChI Key VNGOYPQMJFJDLV-UHFFFAOYSA-N
Synonym dimethyl isophthalate, dimethyl m-phthalate, isophthalic acid dimethyl ester, methyl isophthalate, 1,3-benzenedicarboxylic acid, dimethyl ester, morflex 1129, dimethyl 1,3-benzenedicarboxylate, dimethylisophthalate, methyl 3-carbomethoxy benzoate, isophthalic acid, dimethyl ester
PubChem CID 15088
IUPAC Name dimethyl benzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC=C1)C(=O)OC

Specifications

Melting Point >66.0°C
Color White
Boiling Point 124°C (16.0 hPa)
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4(CO2CH3)2
Quantity 2.5 kg
Solubility Information Solubility in water: insoluble
Viscosity 2.14 mPa.s (99°C)
Formula Weight 194.19
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Dimethyl isophthalate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.