missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl iminodiacetate hydrochloride, 98%

Product Code. 11329939
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11329939 5 g 5g
11339939 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11329939

Brand: Thermo Scientific Alfa Aesar B25729.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39987-25-2 | C6H12ClNO4 | 197.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39987-25-2
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.62
MDL Number MFCD00216724
InChI Key IIWYYIACSUPJCN-UHFFFAOYSA-N
Synonym dimethyl iminodiacetate hydrochloride, dimethyl 2,2'-azanediyldiacetate hydrochloride, methyl 2-2-methoxy-2-oxoethyl amino acetate hydrochloride, dimethyl iminodiacetic acid hydrochloride, iminodiacetic acid dimethyl ester hydrochloride, glycine, n-2-methoxy-2-oxoethyl-, methyl ester, hydrochloride, methyl 2-methoxycarbonyl methyl amino acetate, chloride, zlchem 797, acmc-1aeyr, iminodiacetic dimethyl ester hydrochloride
PubChem CID 2727727
IUPAC Name methyl 2-[(2-methoxy-2-oxoethyl)amino]acetate;hydrochloride
SMILES [H+].[Cl-].COC(=O)CNCC(=O)OC

Specifications

Melting Point ∼170°C
Quantity 5 g
Beilstein 3703236
Formula Weight 197.62
Percent Purity 98%
Chemical Name or Material Dimethyl iminodiacetate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.