missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Heptanedioate-2,2,6,6-d4, CDN
Specifications
Specifications
| Chemical Name or Material | Dimethyl Heptanedioate-2,2,6,6-D4 |
| CAS | 120811-82-7 |
| Molecular Formula | C9 D4 H12 O4 |
| Quantity | 0.5 g |
| Synonym | Heptanedioic-2,2,6,6-d4 acid, dimethyl ester (9 CI), Heptanedioic acid dimethyl ester-2,2,6,6-d4, Dimethyl 1,7-heptanedioate-2,2,6,6-d4, Pimelic acid-2,2,6,6-d4 dimethyl ester, Dimethyl heptanedioate-2,2,6,6-D4, Dimethyl pimelate-2,2,6,6-d4 |
| InChI Formula | InChI=1 S/C9H16O4/c1-12-8(10)6-4-3-5-7-9(11)13-2/h3-7H2,1-2H3/i6D2,7D2 |
| SMILES | [2 H]C([2 H])(CCCC([2 H])([2 H])C(=O)OC)C(=O)OC |
| IUPAC Name | dimethyl 2,2,6,6-tetradeuterioheptanedioate |
| Molecular Weight (g/mol) | 192.2456 |
| Formula Weight | 192.13 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?