missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Fumarate, TRC

Product Code. 17903435
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903435 50 mg 50mg
17984775 100 mg 100mg
17992045 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17903435 Supplier Toronto Research Chemicals Supplier No. TRCD46496550MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-49-7
Molecular Formula C6 H8 O4
Molecular Weight (g/mol) 144.13
IUPAC Name dimethyl (E)-but-2-enedioate
SMILES COC(=O)\C=C\C(=O)OC

Specifications

Quantity 50 mg
Synonym Dimethyl Fumarate, Fumaric acid dimethyl ester, trans-1,2-Ethylenedicarboxylic Acid Dimethyl Ester, 2-Butenedioic acid (2E)-, 1,4-dimethyl ester (ACI), 2-Butenedioic acid (2E)-, dimethyl ester (9CI), 2-Butenedioic acid (E)-, dimethyl ester (ZCI), Fumaric acid, dimethyl ester (6CI, 8CI), (2E)-2-Butenedioic acid dimethyl ester, (E)-2-Butenedioic acid dimethyl ester, (E)-But-2-enedioic acid dimethyl ester, Allomaleic acid dimethyl ester, AZL-O 211089, BG 00012, BG 12, Boletic acid dimethyl ester, Dimethyl (E)-2-butenedioate, Dimethyl (E)-but-2-enedioate, Dimethyl (E)-butenedioate, Dimethyl trans-ethylenedicarboxylate, Marclerosis, Methyl fumarate, NSC 167432, NSC 25942, Panaclar, Skilarence, Tecfidera, Tecfira, trans-Butenedioic acid dimethyl ester
InChI Formula InChI=1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+
Formula Weight 144.0423
Percent Purity >95%
Chemical Name or Material Fumaric acid dimethyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.