missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl fumarate, 99%, Thermo Scientific Chemicals
61.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
250 g
Description
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing
Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).
Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing
Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).
Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
Specifications
Melting Point | 102°C to 105°C |
Density | 1.37 |
Boiling Point | 192°C to 193°C |
Quantity | 250 g |
Beilstein | 774590 |
Merck Index | 14,4287 |
Solubility Information | Soluble in water (1.6mg/ml at 20°C),methanol (30-36mg/ml),ethanol (10mg/ml at 25°C),DMSO (29mg/ml at 25°C),and DMF (∽12mg/ml). |
Formula Weight | 144.13 |
Percent Purity | 99% |
Chemical Name or Material | Dimethyl fumarate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'