missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl fumarate, 99%

Product Code. 11334489
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334489 100 g 100g
11324489 25 g 25g
11408593 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334489 Supplier Thermo Scientific Alfa Aesar Supplier No. A10402.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-49-7 | C6H8O4 | 144.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).

Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-49-7
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00064438
InChI Key LDCRTTXIJACKKU-ONEGZZNKSA-N
Synonym dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate
PubChem CID 637568
ChEBI CHEBI:76004
IUPAC Name dimethyl (E)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point 102°C to 105°C
Density 1.37
Boiling Point 192°C to 193°C
Quantity 100 g
Beilstein 774590
Merck Index 14,4287
Solubility Information Soluble in water (1.6mg/ml at 20°C),methanol (30-36mg/ml),ethanol (10mg/ml at 25°C),DMSO (29mg/ml at 25°C),and DMF (∽12mg/ml).
Formula Weight 144.13
Percent Purity 99%
Chemical Name or Material Dimethyl fumarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.