missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Dodecanedioate, TRC
Specifications
Specifications
| Chemical Name or Material | Dimethyl Dodecanedioate |
| CAS | 1731-79-9 |
| Molecular Formula | C14 H26 O4 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C14H26O4/c1-17-13(15)11-9-7-5-3-4-6-8-10-12-14(16)18-2/h3-12H2,1-2H3 |
| SMILES | COC(=O)CCCCCCCCCCC(=O)OC |
| IUPAC Name | dimethyl dodecanedioate |
| Molecular Weight (g/mol) | 258.35 |
| Formula Weight | 258.1831 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?