missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl disulfide, 99%

Product Code. 11322207
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322207 250 g 250g
11332207 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322207 Supplier Thermo Scientific Alfa Aesar Supplier No. L09679.30
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-92-0 | C2H6S2 | 94.19 g/mol

Dimethyl disulfide is used as an intermediate as well as a food additive in onion, garlic, cheese, meats, soups, savory flavors and fruit flavors. It is used in oil refineries as a sulfiding agent. It is used to alfa substitute 2-methylfuran-acrolein to produce food stuff. It is involved in the preparation of 4-(methylthio)phenol. Further, it is used to replace methyl mercaptan as a jet fuel additive. It finds application in artificial flavoring agent and a corrosion inhibitor. In addition to this, it serves as an effective soil fumigant in agriculture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl disulfide is used as an intermediate as well as a food additive in onion, garlic, cheese, meats, soups, savory flavors and fruit flavors. It is used in oil refineries as a sulfiding agent. It is used to alfa substitute 2-methylfuran-acrolein to produce food stuff. It is involved in the preparation of 4-(methylthio)phenol. Further, it is used to replace methyl mercaptan as a jet fuel additive. It finds application in artificial flavoring agent and a corrosion inhibitor. In addition to this, it serves as an effective soil fumigant in agriculture.

Solubility
Miscible with ethanol, ethyl ether, alcohol, most organic solvents and oils. Slightly miscible with water.

Notes
Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-92-0
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00008561
InChI Key WQOXQRCZOLPYPM-UHFFFAOYSA-N
Synonym dimethyl disulfide, methyl disulfide, dimethyldisulfide, disulfide, dimethyl, dmds, 2,3-dithiabutane, dimethyl disulphide, methyldisulfanyl methane, methyldisulfide, methyldithiomethane
PubChem CID 12232
ChEBI CHEBI:4608
IUPAC Name (methyldisulfanyl)methane
SMILES CSSC

Specifications

Melting Point -85°C
Density 1.046
Boiling Point 108°C to 109°C
Flash Point 10°C (50°F)
Odor Stench
Linear Formula CH3S-SCH3
Refractive Index 1.5253
Quantity 250 g
UN Number UN2381
Beilstein 1730824
Solubility Information Miscible with ethanol,ethyl ether,alcohol,most organic solvents and oils. Slightly miscible with water.
Formula Weight 94.2
Percent Purity 99%
Chemical Name or Material Dimethyl disulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.