missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl diselenide, 99%

Product Code. 11384789
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384789 5 g 5g
11394789 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384789 Supplier Thermo Scientific Alfa Aesar Supplier No. 068119.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7101-31-7 | C2H6Se2 | 188.012 g/mol

Dimethyl diselenide is a powerful reagent used for identification of distonic radical cations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7101-31-7
Molecular Formula C2H6Se2
Molecular Weight (g/mol) 188.012
MDL Number MFCD00008254
InChI Key VLXBWPOEOIIREY-UHFFFAOYSA-N
Synonym dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci
PubChem CID 23496
IUPAC Name (methyldiselanyl)methane
SMILES C[Se][Se]C

Specifications

Density 1.987 g/mL
Boiling Point 155°C to 157°C
Flash Point 60°C (140°F)
Odor Unpleasant
Refractive Index 1.6388
Quantity 5 g
UN Number UN2929
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 187.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl diselenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.