missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Carbonate, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Dimethyl Carbonate |
| CAS | 616-38-6 |
| Molecular Formula | C3 H6 O3 |
| Quantity | 100 g |
| Synonym | Carbonic acid, dimethyl ester, Carbonic acid dimethyl ester, AK 100, AK 100 (carbonate), Dimethyl carbonate, Methyl carbonate, Methyl carbonate ((MeO)2CO), NSC 9371 |
| InChI Formula | InChI=1S/C3H6O3/c1-5-3(4)6-2/h1-2H3 |
| SMILES | COC(=O)OC |
| IUPAC Name | dimethyl carbonate |
| Molecular Weight (g/mol) | 90.0779 |
| Formula Weight | 90.0317 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction