missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl biphenyl-4,4'-dicarboxylate, MedChemExpress

Product Code. 16426036
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16426036 Supplier Medchem Express Supplier No. HY128854
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Dimethyl biphenyl-4,4'-dicarboxylate (Biphenyl dimethyl dicarboxylate) is a hepatoprotectant obtained from Schizandra fructus and may induce a signal transduction similar to that associated with IFN.

Dimethyl biphenyl-4,4'-dicarboxylate (Biphenyl dimethyl dicarboxylate) is a hepatoprotectant obtained from Schizandra fructus and may induce a signal transduction similar to that associated with IFN.

Specifications

Chemical Name or Material Dimethyl biphenyl-4,4'-dicarboxylate
CAS 792-74-5
Color White
Purity Grade Notes Research
Molecular Formula C16H14O4
Quantity 100 mg
Solubility Information DMF : 2 mg/mL (7.40 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (7.40 mM; ultrasonic and heat to 60°C)
SMILES COC(C1=CC=C(C2=CC=C(C(OC)=O)C=C2)C=C1)=O
Molecular Weight (g/mol) 270.28
Formula Weight 270.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.