missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl biphenyl-4,4'-dicarboxylate, 98%, Thermo Scientific™

Product Code. 11424254
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11424254

Brand: Thermo Scientific Alfa Aesar A12556.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Dimethyl biphenyl-4,4′-dicarboxylate is used as a hepatoprotectant in the treatment of chronic hepatitis. At 250 µg/ml, it has been shown to stimulate JAK/STAT signaling and to induce the expression of interferon-α stimulated genes in a HepG2 cell line. It has been used to study hepatic fibrosis and chronic hepatocellular damage. It appears to normalize alanine aminotransferase (ALT) in cases where levels of ALT are elevated. It may be employed as starting reagent for the synthesis of dimethyl 2-fluoro-and 2,2′-difluorobiphenyl-4,4′-dicarboxylates.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 792-74-5
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.28
MDL Number MFCD00017201
InChI Key BKRIRZXWWALTPU-UHFFFAOYSA-N
Synonym dimethyl 1,1'-biphenyl-4,4'-dicarboxylate, dimethyl biphenyl-4,4'-dicarboxylate, biphenyl dimethyl dicarboxylate, dimethyl 4,4'-biphenyldicarboxylate, 1,1'-biphenyl-4,4'-dicarboxylic acid, dimethyl ester, unii-k61bxa0u9c, 4,4'-biphenyldicarboxylic acid, dimethyl ester, k61bxa0u9c, methyl 4-4-methoxycarbonylphenyl benzoate, methyl 4-4-methoxycarbonyl phenyl benzoate
PubChem CID 13099
IUPAC Name methyl 4-(4-methoxycarbonylphenyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 212°C to 216°C
Quantity 1 g
Beilstein 2055852
Solubility Information Insoluble in water.
Formula Weight 270.29
Percent Purity 98%
Chemical Name or Material Dimethyl biphenyl-4,4'-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.