missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Aminoterephthalate, TRC

Product Code. 30381837
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381837 100 g
30386430 25 g
30379849 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381837 Supplier Toronto Research Chemicals Supplier No. TRCD460350100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5372-81-6
Molecular Formula C10 H11 N O4
Molecular Weight (g/mol) 209.19864
Synonym Aminoterephthalic acid dimethyl ester, Dimethyl 2-aminobenzene-1,4-dicarboxylate, Dimethyl 2-aminobenzene-1,4-dioate, Dimethyl 2-aminoterephthalate, Dimethyl 3-aminoterephthalate, Dimethyl o-aminoterephthalate, NSC 20561
IUPAC Name dimethyl 2-aminobenzene-1,4-dicarboxylate
SMILES O=C(OC)C1=CC=C(C(=O)OC)C(N)=C1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H11NO4/c1-14-9(12)6-3-4-7(8(11)5-6)10(13)15-2/h3-5H,11H2,1-2H3
Formula Weight 209.0688
Chemical Name or Material Dimethyl Aminoterephthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.