missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl allylphosphonate, tech. 85%, Thermo Scientific Chemicals

Product Code. 11328515
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328515 5 g 5g
11318515 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328515 Supplier Thermo Scientific Alfa Aesar Supplier No. L10567.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 757-54-0
Molecular Formula C5H11O3P
Molecular Weight (g/mol) 150.114
MDL Number MFCD00014951
InChI Key ZOSQAGGCVFVCNO-UHFFFAOYSA-N
Synonym dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester
PubChem CID 136570
IUPAC Name 3-dimethoxyphosphorylprop-1-ene
SMILES COP(=O)(CC=C)OC

Specifications

Boiling Point 41°C to 42°C (0.5 mmHg)
Refractive Index 1.437
Quantity 5 g
Beilstein 1756526
Formula Weight 150.12
Percent Purity 85%
Grade Technical
Chemical Name or Material Dimethyl allylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.