missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl allylphosphonate, 97%, ACROS Organics™

Product Code. 10514151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10514151 1 g Glass bottle 1g
10489281 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10514151 Supplier Thermo Scientific Acros Supplier No. 330680010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 757-54-0
Molecular Formula C5H11O3P
Molecular Weight (g/mol) 150.114
MDL Number MFCD00014951
InChI Key ZOSQAGGCVFVCNO-UHFFFAOYSA-N
Synonym dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester
PubChem CID 136570
IUPAC Name 3-dimethoxyphosphorylprop-1-ene
SMILES COP(=O)(CC=C)OC

Specifications

Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4360 to 1.4400 (20°C, 589nm)
Quantity 1 g
Formula Weight 150.11
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Dimethyl allylphosphonate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.