missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Adipate, TRC
Specifications
Specifications
| Chemical Name or Material | Adipic acid-dimethyl ester |
| CAS | 627-93-0 |
| Molecular Formula | C8 H14 O4 |
| Quantity | 2.5 g |
| Synonym | Adipic acid, dimethyl ester (6CI,8CI), Hexanedioic acid, dimethyl ester (9CI), Adipinic acid dimethyl ester, DBE 6, Dimethyl 1,6-hexanedioate, Dimethyl adipate, Dimethyl hexanedioate, Methyl adipate, NSC 11213 |
| InChI Formula | InChI=1S/C8H14O4/c1-11-7(9)5-3-4-6-8(10)12-2/h3-6H2,1-2H3 |
| SMILES | COC(=O)CCCCC(=O)OC |
| IUPAC Name | dimethyl hexanedioate |
| Molecular Weight (g/mol) | 174.19 |
| Formula Weight | 174.0892 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?