missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl adipate, 99%

Product Code. 11454667
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454667 250 g 250g
11344489 50 g 50g
11444667 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454667 Supplier Thermo Scientific Alfa Aesar Supplier No. B21174.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-93-0 | C8H14O4 | 174.196 g/mol

Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

Solubility
Miscible with alcohols and ether. Immiscible with water.

Notes
Moisture sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-93-0
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00008469
InChI Key UDSFAEKRVUSQDD-UHFFFAOYSA-N
Synonym dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62
PubChem CID 12329
ChEBI CHEBI:34715
IUPAC Name dimethyl hexanedioate
SMILES COC(=O)CCCCC(=O)OC

Specifications

Melting Point 8°C
Density 1.063
Boiling Point 109°C to 110°C (14 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.428
Quantity 250 g
Beilstein 1707443
Merck Index 14,162
Solubility Information Miscible with alcohols and ether. Immiscible with water.
Formula Weight 174.2
Percent Purity 99%
Chemical Name or Material Dimethyl adipate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.