missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylsuccinate, 96%

Product Code. 11447106
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447106 500 g 500g
11437106 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447106 Supplier Thermo Scientific Alfa Aesar Supplier No. B25423.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10420-33-4 | C8H12O5 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10420-33-4
Molecular Formula C8H12O5
Molecular Weight (g/mol) 188.18
MDL Number MFCD00008448
InChI Key XREKLQOUFWBSFH-UHFFFAOYNA-N
Synonym dimethyl acetylsuccinate, dimethyl 2-acetylsuccinate, 1,4-dimethyl 2-acetylbutanedioate, butanedioic acid, acetyl-, dimethyl ester, acetylsuccinic acid dimethyl ester, dmas, butanedioic acid, 2-acetyl-, 1,4-dimethyl ester, doimethyl acetylsuccinate, acmc-1bwle, dimethyl 2-acetylsuccite
PubChem CID 112039
IUPAC Name dimethyl 2-acetylbutanedioate
SMILES COC(=O)CC(C(C)=O)C(=O)OC

Specifications

Melting Point 30°C to 33°C
Density 1.16
Boiling Point 129°C to 134°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 500 g
Formula Weight 188.18
Percent Purity 96%
Chemical Name or Material Dimethyl acetylsuccinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.