missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylenedicarboxylate, 98%

Product Code. 10569250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10569250 100 g 100g
10428260 5 g 5g
10356840 25 g 25g
11925031 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10569250 Supplier Thermo Scientific Acros Supplier No. 115711000
  • ONLINE EXCLUSIVE PRICE
    £151.00
    £135.65 / 100g
    Save £15.35
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 762-42-5 | C6H6O4 | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-42-5
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
InChI Key VHILMKFSCRWWIJ-UHFFFAOYSA-N
Synonym dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene
PubChem CID 12980
IUPAC Name dimethyl but-2-ynedioate
SMILES COC(=O)C#CC(=O)OC

Specifications

Melting Point -18.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 95.0°C to 98.0°C (19.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CC≡CCO2CH3
Refractive Index 1.4460 to 1.4480
Quantity 100 g
Beilstein 02,803
Fieser 01,272; 02,145; 03,103; 04,168; 05,227; 06,206; 07,117; 09,176
Specific Gravity 1.16
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethyl alcohol,ethyl ether,ccl4,miscible with organic solvents
Formula Weight 142.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl acetylenedicarboxylate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl acetylenedicarboxylate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.